3,5-Dinitrosalicylic acid ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:609-99-4
:
Formula:C₇H₄N₂O₇
MDL Number:MFCD00007104
:228.12 g/mol
Melting Pt:169…174 °C
Storage Temperature:Ambient
EINECS:210-204-3
UN:2811
ADR:6.1,III
:GHS07
Related Information
Frequently Bought Together