

Benzoic acid ≥99.5%, certified reference material for titrimetry, Supelco®
Catalog # 12353-25G-F
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:122.12 g/mol
Merck Index:13,01092
Boiling Pt:249 °C (1013 hPa)
Melting Pt:121…125 °C
Density:1.32 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ300
Flash Pt:121 °C (closed cup)
Storage Temperature:Ambient
EINECS:200-618-2
REACH:01-2119455536-33
:GHS05
:GHS08
CAS Number:65-85-0
Formula:C₇H₆O₂
MDL Number:MFCD00002398