4-Bromophenol 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:106-41-2
:
Formula:C₆H₅BrO
MDL Number:MFCD00002313
:173.01 g/mol
Merck Index:13,01413
Boiling Pt:236 °C (1013 hPa)
Melting Pt:64…68 °C
Density:~ 1,84 g/cm³ (20 °C)
Flash Pt:≥ 110 °C
Storage Temperature:Ambient
EINECS:203-394-4
ADR:6.1,III
:GHS07
Specification Test Results
- Appearance:White to pink to light brown Crystalline powder or crystals or solid
- Infrared spectrum:Conforms
- GC:≥96.0 %
Specification Test Results
Frequently Bought Together