


4-Bromoaniline for synthesis, Sigma-Aldrich®
Catalog # 8.01600.0100
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 g
- Seq:3401
Specifications
Specification Test Results
- Solubility:(20 °C) practically insoluble
- Melting Point:60 - 64 °C
- Molar Mass:172.02 g/mol
- Bulk Density:810 kg/m3
- Boiling Point:230 - 250 °C (1013 hPa)
Specification Test Results
About this item
:
Merck Index:13,01388
Boiling Pt:223 °C (1013 hPa)
Melting Pt:62…64 °C
Density:1.799 g/cm³ (20 °C)
ADR:9,III
:M-Clarity™ quality level = MQ200
:172.02 g/mol
EINECS:203-393-9
UN:3077
:GHS06
:GHS08
:GHS09
Storage Temperature:Ambient
CAS Number:106-40-1
Formula:C₆H₆BrN
MDL Number:MFCD00007822