Supelco® Organic Reference Standard, Heptadecafluorooctanesulfonic Acid Solution
Catalog # 33607-1ML
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- Standards:Analytical standard
- CAS No.:1763-23-1
- Description:Heptadecafluorooctanesulfonic acid solution
- Storage conditions:2...8 °C
- Hazard class description:H225 - H301 + H311 + H331 - H370
- Shipping conditions:STD
- MDL No.:MFCD00042454
- M-Clarity quality level:MQ100
- Synonym(s):PFAS; PFOS; Perfluorooctanesulfonic acid
- Formula:CF₃(CF₂)₇SO₃H
- Molecular weight:500,13
- Flash point:11 °C - closed cup
Specifications
About this item
- GHS signal: GHS02, GHS06, GHS08
Caution: P210 - P233 - P280 - P301 + P310 - P303 + P361 + P353 - P304 + P340 + P311