Order Entry
Slovakia
ContactUsLinkComponent
Supelco® Organic Reference Standard, Pentadecafluorooctanoic Acid Solution
Supelco® Organic Reference Standard, Pentadecafluorooctanoic Acid Solution
Catalog # 33603-1ML
Supplier:  Merck
CAS Number:  
undefined
Supelco® Organic Reference Standard, Pentadecafluorooctanoic Acid Solution
Catalog # 33603-1ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • Standards:
    Analytical standard
  • CAS No.:
    335-67-1
  • Description:
    Pentadecafluorooctanoic acid solution
  • Storage conditions:
    2...8 °C
  • Hazard class description:
    H225 - H301 + H311 + H331 - H370
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00004174
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    PFAS; PFOA; Perfluorocaprylic acid solution; Perfluorooctanoic acid solution
  • Formula:
    CF₃(CF₂)₆COOH
  • Molecular weight:
    414,07
  • Flash point:
    11 °C - closed cup

Specifications

About this item

  • GHS signal: GHS02, GHS06, GHS08
Caution: P210 - P233 - P280 - P301 + P310 - P303 + P361 + P353 - P304 + P340 + P311