Order Entry
Slovakia
ContactUsLinkComponent
Organic reference standard, 1,3-Diethylbenzene 100 µg/ml in methanol, Dr Ehrenstorfer
Organic reference standard, 1,3-Diethylbenzene 100 µg/ml in methanol, Dr Ehrenstorfer
Catalog # EHERXA12605500ME
Supplier:  EHRENSTORFER
CAS Number:  
Organic reference standard, 1,3-Diethylbenzene 100 µg/ml in methanol, Dr Ehrenstorfer
Catalog # EHERXA12605500ME
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    141-93-5
  • Description:
    1,3-Diethylbenzene 100 µg/ml in methanol
  • Formula:
    C₁₀H₁₄
  • Description:
    1,3-Diethylbenzene

Specifications

About this item