Order Entry
Slovakia
ContactUsLinkComponent
Toluenediisocyanate ≥99% (2,4-80%, 2,6-20%)
Toxic
Health
Toluenediisocyanate ≥99% (2,4-80%, 2,6-20%)
Catalog # ABCRAB142608.0500
Supplier:  abcr
CAS Number:  
Toxic
Health
Toluenediisocyanate ≥99% (2,4-80%, 2,6-20%)
Catalog # ABCRAB142608.0500
Supplier:  abcr
CAS Number:  

Specifications

  • Pk:
    500 g

Specifications

About this item

 :
ADR:6.1,II
 :174.16
EINECS:247-722-4
UN:2078
 :GHS06
 :GHS08
Storage Temperature:Ambient
CAS Number:26471-62-5
 :584-84-9
Formula:C₉H₆N₂O₂
MDL Number:MFCD01778251