Supelco® Organic Reference Standard, 1-Decene
Supplier: Merck
Certificates
About this item
- GHS signal: GHS02, GHS08, GHS09
Caution: P210 - P233 - P240 - P273 - P301 + P310 - P331
Specifications
- CAS No.:872-05-9
- Storage conditions:2...30 °C
- Hazard class description:H226 - H304 - H410
- Shipping conditions:STD
- MDL No.:MFCD00009577
- M-Clarity quality level:MQ100
- Melting point:–66,3…+66 °C (lit.)
- Formula:CH₃(CH₂)₇CH=CH₂
- Molecular weight:140,27
- Density:0,741 g/ml at 25 °C (lit.)
- Flash point:49 °C
- Boiling point:166,5...173,5 °C (lit.)
Specifications
Frequently Bought Together