Order Entry
Slovakia
ContactUsLinkComponent
Supelco® Organic Reference Standard, Tetrachloroethylene
Supelco® Organic Reference Standard, Tetrachloroethylene
Supplier:  Merck
Certificates
undefined
Supelco® Organic Reference Standard, Tetrachloroethylene
Supplier:  Merck
Certificates

Specifications

  • CAS No.:
    127-18-4
  • Storage conditions:
    RT
  • Hazard class description:
    H315 - H317 - H319 - H336 - H351 - H411
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00000834
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    PCE; Perchloroethylene
  • Melting point:
    –22 °C (lit.)
  • Formula:
    CCl₂=CCl₂
  • Molecular weight:
    165,83
  • Density:
    1,623 g/ml at 25 °C (lit.)
  • Boiling point:
    121 °C (lit.)

Specifications

Related Information

Frequently Bought Together