Supelco® Organic Reference Standard, 2,2,4-Trimethylpentane
Supplier: Merck
Certificates
About this item
- GHS signal: GHS02, GHS08, GHS07, GHS09
Caution: P210 - P273 - P301 + P310 + P331 - P302 + P352
Specifications
- CAS No.:540-84-1
- Storage conditions:2...30 °C
- Hazard class description:H225 - H304 - H315 - H336 - H410
- Shipping conditions:STD
- MDL No.:MFCD00008943
- M-Clarity quality level:MQ100
- Synonym(s):Isooctane
- Melting point:–107 °C (lit.)
- Formula:(CH₃)₂CHCH₂C(CH₃)₃
- Molecular weight:114,23
- Density:0,692 g/ml at 25 °C (lit.)
- Flash point:–12 °C - closed cup
- Boiling point:98...99 °C (lit.)
Specifications
Frequently Bought Together