Order Entry
Slovakia
ContactUsLinkComponent
Supelco® Organic Reference Standard, Tricosane
Supelco® Organic Reference Standard, Tricosane
Supplier:  Merck
Certificates
undefined
Supelco® Organic Reference Standard, Tricosane
Supplier:  Merck
Certificates

Specifications

  • CAS No.:
    638-67-5
  • Storage conditions:
    RT
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00009350
  • M-Clarity quality level:
    MQ100
  • Melting point:
    46 - 47 °C (lit.); 47 - 49 °C
  • Formula:
    CH₃(CH₂)₂₁CH₃
  • Molecular weight:
    324,63
  • Flash point:
    113 °C - Closed cup
  • Boiling point:
    199 - 200 °C/3 mmHg (lit.)

Specifications

Related Information

Frequently Bought Together