n-Octadecane, Supelco®
Catalog # 74691-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:254.5 g/mol
Boiling Pt:316.1 °C (1013 hPa)
Melting Pt:26…29 °C
Density:0.7765 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
Flash Pt:165 °C
Storage Temperature:Ambient
EINECS:209-790-3
CAS Number:593-45-3
Formula:H₃C(CH₂)₁₆CH₃
MDL Number:MFCD00009007