n-Nonacosane, Supelco®
Catalog # 74156-1G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:408.8 g/mol
Boiling Pt:440.8 °C (1013 hPa)
Melting Pt:63.7 °C
Density:0.8083 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:211-126-2
CAS Number:630-03-5
Formula:H₃C(CH₂)₂₇CH₃
MDL Number:MFCD00009356