

1-Pentanol ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:71-41-0
:
Formula:H₃C(CH₂)₃CH₂OH
MDL Number:MFCD00002977
:88.15 g/mol
Merck Index:14,7118
Boiling Pt:136…138 °C
Melting Pt:–79 °C
Density:0.812
Flash Pt:49 °C (120 °F)
Storage Temperature:Ambient
EINECS:200-752-1
UN:1105
ADR:3,III
:GHS02
:GHS07
Related Information
Frequently Bought Together