Triethylamine ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:121-44-8
:
Formula:(C₂H₅)₃N
MDL Number:MFCD00009051
:101.19 g/mol
Merck Index:14,9666
Boiling Pt:89…90 °C
Melting Pt:–115 °C
Density:0.726
Flash Pt:–11 °C (–12 °F)
Storage Temperature:Ambient
EINECS:204-469-4
UN:1296
ADR:3,II
:GHS02
:GHS05
:GHS07
Related Information
Frequently Bought Together