Order Entry
Slovakia
ContactUsLinkComponent
Citric acid, anhydrous ≥96%, grit, tested according to Ph. Eur., Fluka™
Citric acid, anhydrous ≥96%, grit, tested according to Ph. Eur., Fluka™
Catalog # 03878-1KG
Supplier:  HONEYWELL
CAS Number:  
undefined
Citric acid, anhydrous ≥96%, grit, tested according to Ph. Eur., Fluka™
Catalog # 03878-1KG
Supplier:  HONEYWELL
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

Specification Test Results

  • pKa :
    (1) 3.13, (2) 4.76, (3) 6.4
  • Identity (IR):
    complying
  • Assay (calculated on the anhydrous basis):
    99.5 - 100.5 %
  • Appearance of the solution:
    complying
  • Sulfated ash:
    Max. 0.1 %
  • Water (Karl Fischer):
    Max. 1.0 %
  • Oxalate (as Oxalic acid):
    Max. 360 ppm
  • Heavy metals (as Pb):
    Max. 10 ppm
  • Sulfate (SO₄):
    Max. 150 ppm
  • Reaction against H₂SO₄:
    complying
  • Residual solvents:
    complying
  • calcium (Ca):
    Max. 0.005 %
  • cobalt (Co):
    Max. 0.002 %
  • copper (Cu):
    Max. 0.005 %
  • iron (Fe):
    Max. 0.001 %
  • potassium (K):
    Max. 0.01 %
  • magnesium (Mg):
    Max. 0.005 %
  • manganese (Mn):
    Max. 0.002 %
  • sodium (Na):
    Max. 0.05 %
  • vanadium (V):
    Max. 0.001 %
  • zirconium (Zr):
    Max. 0.001 %
  • chloride (Cl):
    Max. 0.001 %
  • total N:
    Max. 0.002 %

Specification Test Results

About this item

 :
 :192.13 g/mol
Merck Index:13,02350
Boiling Pt:310 °C (1013 hPa)
Melting Pt:153-159 °C
Melting Pt:153…159 °C
Density:1.67 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:201-069-1
REACH:01-2119457026-42-0000
 :GHS07
CAS Number:77-92-9
Formula:HOC(COOH)(CH₂COOH)₂
MDL Number:MFCD00011669