Order Entry
Slovakia
ContactUsLinkComponent
Undecanoic acid
Undecanoic acid
Catalog # EHERC17896400
Supplier:  EHRENSTORFER
CAS Number:  
Undecanoic acid
Catalog # EHERC17896400
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,25 g
  • Seq:
    3029

Specifications

About this item

 :
 :186.29 g/mol
Boiling Pt:228 °C (1013 hPa)
Melting Pt:30 °C
Density:0.89 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:203-964-2
CAS Number:112-37-8
Formula:C₁₁H₂₂O₂
MDL Number:MFCD00002730