


Formic acid ≥94.5%, high purity
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:64-18-6
:
Formula:HCO₂H
MDL Number:MFCD00003297
:46.03 g/mol
Boiling Pt:100…101 °C (1013 hPa)
Melting Pt:8.2…8.4 °C
Density:1.22 g/cm³ (25 °C)
Flash Pt:48 °C
Storage Temperature:Ambient
EINECS:200-579-1
UN:1779
ADR:8 (3),II
REACH:01-2119491174-37
:GHS02
:GHS05
:GHS06
Specification Test Results
- Purity:≥94.5%
Specification Test Results
Related Information
Certificates
Search by Lot NumberVWR® for chromatography columns, consumables and reagents
Product Literature
Frequently Bought Together