Order Entry
Slovakia
ContactUsLinkComponent
Cerotic acid
  EHERC14191300
 :  
Cerotic acid
  EHERC14191300
 :  
  • Pack type:
    Glass bottle for solids
  • Pk:
    20 mg
  • Seq:
    2305
 :
 :396.7 g/mol
 :419 °C (1013 hPa)
 :76…77 °C
 :0,8198 g/cm³ (100 °C)
 :Refrigerator
 :208-040-2
 :506-46-7
 :C₂₆H₅₂O₂
 :MFCD00002811