Order Entry
Slovakia
Orders LinkContactUsLinkComponent
 

Hexyl acetate, Supelco®

Glass bottle, 1 mL
Catalog # 25539-1ML
Environment
Supplier:   Merck
Hexyl acetate, Supelco®
Catalog # 25539-1ML
Supplier:  Merck
CAS Number:  142-92-7
Hexyl acetate, Supelco®
Hexyl acetate, Supelco®

Glass bottle, 1 mL

CAS Number:  142-92-7
Price
36,58 €
Log in for availability
1 * 1 mL
Product Details & Documents
Synonyms:  Acetic acid hexyl ester
  • CAS Number:142-92-7
  • Formula:C₈H₁₆O₂
  • MDL Number:MFCD00009524
  • Molecular Weight:144.21 g/mol
  • Boiling Pt:338 °C (1013 hPa)
  • Melting Pt:–80 °C
  • Density:0.87 g/cm³ (20 °C)
  • Flash Pt:56 °C
  • Storage Temperature:Refrigerator
  • EINECS:205-572-7
  • ADR:3,III
  •  :M-Clarity™ quality level = MQ100
  • UN:3271
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL

Specifications