Supelco® Organic Reference Standard, Ethyl acetate
Catalog # 58958-5ML
Supplier: Merck
CAS Number:
Specifications
- Pk:5 mL
- Standards:Analytical standard
- CAS No.:141-78-6
- Description:Ethyl acetate
- Seq:2210
- Storage conditions:RT
- Hazard class description:H225 - H319 - H336
- Shipping conditions:STD
- MDL No.:MFCD00009171
- M-Clarity quality level:MQ200
- Synonym(s):EtOAc
- Melting point:–84 °C (lit.)
- Formula:CH₃COOC₂H₅
- Molecular weight:88,11
- Density:0,902 g/ml at 25 °C (lit.)
- Flash point:–4 °C
- Boiling point:76.5 - 77.5 °C (lit.)
Specifications
About this item
- GHS signal: GHS02, GHS07
Caution: P210 - P233 - P240 - P241 - P242 - P305 + P351 + P338