5-Nonanone ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:502-56-7
:
Formula:C₉H₁₈O
MDL Number:MFCD00009484
:142.24 g/mol
Boiling Pt:186…187 °C
Melting Pt:–50 °C
Density:0.824
Flash Pt:65 °C (149 °F)
Storage Temperature:Ambient
EINECS:207-946-5
Related Information
Frequently Bought Together