Order Entry
Slovakia
ContactUsLinkComponent
3-Pentanone
3-Pentanone
  EHERC15981600
 :  
3-Pentanone
  EHERC15981600
 :  
  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • Seq:
    2070
 :
 :86.13 g/mol
 :13,03148
 :101 °C (1013 hPa)
 :–40 °C
 :0.8148 g/cm³ (20 °C)
 :13 °C
 :Ambient
 :202-490-3
 :1156
 :3,II
 :GHS02
 :GHS07
 :96-22-0
 :C₅H₁₀O
 :MFCD00009320