

3-Pentanone 98%, pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:96-22-0
:
Formula:C₅H₁₀O
MDL Number:MFCD00009320
:86.13 g/mol
Merck Index:13,03148
Boiling Pt:100…102 °C (760 mmHg)
Melting Pt:–40 °C
Density:0.815 g/cm³
Flash Pt:7 °C
Storage Temperature:Ambient
EINECS:202-490-3
UN:1156
ADR:3,II
:GHS07
:GHS02
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.3910 to 1.3930 (20°C, 589 nm)
- GC:≥97.5 %
Specification Test Results
Frequently Bought Together