Order Entry
Slovakia
Orders LinkContactUsLinkComponent
 

Coronene

Glass bottle for solids, 20 mg
Catalog # PROOBCR-272
Supplier:   PROMOCHEM
Coronene
Catalog # PROOBCR-272
Supplier:  PROMOCHEM
CAS Number:  191-07-1
Coronene

Glass bottle for solids, 20 mg

CAS Number:  191-07-1
Price
419,65 €
Log in for availability
1 * 20 mg
Product Details & Documents
Synonyms:  Hexabenzobenzene
  • CAS Number:191-07-1
  • Formula:C₂₄H₁₂
  • MDL Number:MFCD00004134
  • Molecular Weight:300.36 g/mol
  • Boiling Pt:525 °C (1013 hPa)
  • Melting Pt:437…440 °C
  • Density:1.371 g/cm³ (25 °C)
  • EINECS:205-881-7
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    20 mg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.