Supelco® PESTANAL® Organic Reference Standard, Bromoxynil
Catalog # 45355-250MG
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:250 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:1689-84-5
- Description:Bromoxynil
- Storage conditions:2...30 °C
- Hazard class description:H301 - H317 - H330 - H361d - H410
- Shipping conditions:STD
- MDL No.:MFCD00002153
- M-Clarity quality level:MQ100
- Melting point:189...191 °C (lit.)
- Formula:Br₂C₆H₂(OH)CN
- Molecular weight:276,91
- Flash point:Not applicable
Specifications
About this item
- GHS signal: GHS06, GHS08, GHS09
Caution: P202 - P260 - P273 - P280 - P302 + P352 - P304 + P340 + P310