Order Entry
Slovakia
ContactUsLinkComponent
1-Butanesulphonic acid sodium salt, LiChropur™ for ion pair chromatography, Supelco®
1-Butanesulphonic acid sodium salt, LiChropur™ for ion pair chromatography, Supelco®
Catalog # 1.18303.0025
Supplier:  Merck
CAS Number:  
1-Butanesulphonic acid sodium salt, LiChropur™ for ion pair chromatography, Supelco®
Catalog # 1.18303.0025
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Hill Formula:
    C₄H₉NaO₃S
  • Molar Mass:
    160.17 g/mol
  • Melting Point:
    310 °C
  • Storage class:
    10 - 13 Other liquids and solids
  • WGK:
    WGK 3 highly water endangering

Specification Test Results

About this item

 :
 :160.17 g/mol
Melting Pt:> 300 °C
 :M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:219-201-1
CAS Number:2386-54-1
Formula:C₄H₉NaO₃S
MDL Number:MFCD00007540