
Trimethyl orthobenzoate ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:707-07-3
:
Formula:C₁₀H₁₄O₃
MDL Number:MFCD00008474
:182.22 g/mol
Boiling Pt:87…88 °C (7 mmHg)
Density:1.060
Flash Pt:82 °C (179 °F)
EINECS:211-897-5
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together