Acetonitrile 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Methyl cyanide, Ethyl nitrile, Cyanomethane, Ethanenitrile, Ethanoic acid nitrile
CAS Number:75-05-8
:
Formula:H₃CCN
MDL Number:MFCD00001878
Boiling Pt:81…82 °C (760 mmHg)
Melting Pt:–46 °C
Density:0.781 g/cm³
Molecular Weight:41.05 g/mol
Flash Pt:2 °C
Storage Temperature:Ambient
UN:1648
ADR:3 II
Merck Index:13,00071
EINECS:200-835-2
REACH:01-2119471307-38
:GHS07
:GHS02
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Clear liquid
- Water:≤0.03 %
- GC:≥99.0 %
- UV:at 193 nm A: ≤1
- Color scale:≤10 APHA
- Residue after evaporation:≤0.0003 %
Specification Test Results
Frequently Bought Together