
Magnesium nitrate hexahydrate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:13446-18-9
:10213-15-7
Formula:Mg(NO₃)₂·6H₂O
MDL Number:MFCD00149779
:256.41 g/mol
Merck Index:13,05697
Boiling Pt:330 °C (1013 hPa)
Melting Pt:89 °C
Density:1.63 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:233-826-7
UN:1474
ADR:5.1,III
:M-Clarity™ quality level = MQ200
Flash Pt:93.3 °C
:GHS03
Frequently Bought Together