Barium chloride dihydrate ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10326-27-9
:
Formula:BaCl₂·2H₂O
MDL Number:MFCD00149154
:244.27 g/mol
Merck Index:14,971
Boiling Pt:1560 °C (1013 hPa)
Melting Pt:113 °C (-2H₂O)
Density:3.097
Storage Temperature:Ambient
EINECS:233-788-1
UN:1564
ADR:6.1,III
:GHS06
Related Information
Frequently Bought Together