Sodium tungstate dihydrate ≥99%, Purum, Fluka™
14304-6X500G
:
- Pack type:Plastic bottle
- Pk:3.000 g
- Seq:3810
- Environmentally Preferable:
- Assay:Min. 99 %
- Free alkali (as NaOH):Max. 0.1 %
- Iron (Fe):Max. 0.002 %
- Molybdenum (Mo):Max. 0.01 %
- Chloride (Cl):Max. 0.01 %
- Sulfate (SO₄):Max. 0.02 %
: Tungstic acid sodium salt dihydrate, Sodium wolframate dihydrate, Sodium tungsten oxide dihydrate
:
:329.85 g/mol
:698 °C
:236-743-4
:GHS07
:Ambient
:10213-10-2
:Na₂WO₄·2H₂O
:MFCD00149190
:4.18 g/cm³
:12,08847