Order Entry
Slovakia
ContactUsLinkComponent
Supelco® Organic Reference Standard, (−)-Camphor
Supelco® Organic Reference Standard, (−)-Camphor
Catalog # 21293-1G
Supplier:  Merck
CAS Number:  
undefined
Supelco® Organic Reference Standard, (−)-Camphor
Catalog # 21293-1G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Standards:
    Analytical standard
  • CAS No.:
    464-48-2
  • Description:
    (−)-Camphor
  • Storage conditions:
    2...30 °C
  • Hazard class description:
    H228 - H315 - H319 - H335
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00064148
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    (1S)-1,7,7-Trimethylbicyclo[2.2.1]heptan-2-one
  • Molecular weight:
    152,23
  • Flash point:
    65 °C - closed cup

Specifications

About this item

  • GHS signal: GHS02, GHS07
Caution: P210 - P302 + P352 - P305 + P351 + P338