(±)-2-Butanol, Sigma-Aldrich®
: Merck
:78-92-2
:
:H₃CCH₂CH(OH)CH₃
:MFCD00004569
:74.12 g/mol
:13,01540
:94 °C (1013 hPa)
:–115 °C
:0.8534 g/cm³ (20 °C)
:201-158-5
:1120
:3,III
:M-Clarity™ quality level = MQ100
:23 °C
:GHS02
:GHS07
:Ambient
Frequently Bought Together