Order Entry
Slovakia
ContactUsLinkComponent
Hydrofluoric acid 40%, Sigma-Aldrich®
Toxic
Corrosive
Hydrofluoric acid 40%, Sigma-Aldrich®
Catalog # SIAL30112-60KG-H
Supplier:  Merck
CAS Number:  
Toxic
Corrosive
Hydrofluoric acid 40%, Sigma-Aldrich®
Catalog # SIAL30112-60KG-H
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    60 kg
  • Seq:
    4463

Specifications

About this item

 :
 :20.01 g/mol
Storage Temperature:Ambient
EINECS:231-634-8
UN:1790
ADR:8,II
 :GHS06
 :GHS05
CAS Number:7664-39-3
Formula:HF
MDL Number:MFCD00011346