


Formic acid ≥88% ACS
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:64-18-6
:
Formula:HCO₂H
MDL Number:MFCD00003297
:46.03 g/mol
Merck Index:14,4241
Boiling Pt:100…101 °C
Melting Pt:7…9 °C
Density:1.220
Flash Pt:50 °C (156 °F)
Storage Temperature:Ambient
EINECS:200-579-1
UN:1779
ADR:8,II
:GHS06
:GHS05
:GHS02
Related Information
Frequently Bought Together