Supelco® Organic Reference Standard, Capsaicin
Supplier: Merck
Certificates
About this item
- GHS signal: GHS06, GHS08, GHS05
Caution: P261 - P280 - P301 + P310 - P302 + P352 - P304 + P340 + P312 - P305 + P351 + P338
Specifications
- CAS No.:404-86-4
- Storage conditions:2...8 °C
- Hazard class description:H300 - H315 - H317 - H318 - H334 - H335
- Shipping conditions:STD
- MDL No.:MFCD00017259
- M-Clarity quality level:MQ100
- Synonym(s):8-Methyl-N-vanillyl-trans-6-nonenamide
- Melting point:62...65 °C (lit.)
- Formula:(CH₃)₂CHCH=CH(CH₂)₄CONHCH₂C₆H₃-4-(OH)-3-(OCH₃)
- Molecular weight:305,41
- Flash point:113 °C - closed cup
Specifications
Frequently Bought Together