Order Entry
Slovakia
ContactUsLinkComponent
Supelco® Organic Reference Standard, (+)-β-Pinene
Supelco® Organic Reference Standard, (+)-β-Pinene
Supplier:  Merck
Certificates
undefined
Supelco® Organic Reference Standard, (+)-β-Pinene
Supplier:  Merck

Specifications

  • CAS No.:
    19902-08-0
  • Storage conditions:
    2…8 °C
  • Hazard class description:
    H226 - H315 - H319 - H335
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00151105
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    (1R,5R)-2(10)-Pinene; (1R,5R)-6,6-Dimethyl-2-methylenebicyclo[3.1.1]heptane
  • Molecular weight:
    136,23
  • Density:
    0,872 g/ml at 25 °C (lit.)
  • Flash point:
    36 °C - Closed cup
  • Boiling point:
    164 - 165 °C (lit.)

Specifications

Frequently Bought Together