Order Entry
Slovakia
ContactUsLinkComponent
Iodine 5 g/l in chloroform Reag. Ph. Eur. 1045805
Iodine 5 g/l in chloroform Reag. Ph. Eur. 1045805
Catalog # 87855.290
Supplier:  VWR Chemicals
CAS Number:  
undefined
Iodine 5 g/l in chloroform Reag. Ph. Eur. 1045805
Catalog # 87855.290
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :253.81 g/mol
Storage Temperature:Ambient
EINECS:231-442-4
CAS Number:7553-56-2
Formula:I₂
MDL Number:MFCD00011355