Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovakia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Electrochemistry Meters
pH and Multi-Parameter Meters
Accessories for Jenway meters
Accessories for Jenway meters
Accessories for Jenway meters
Catalog # JENW99960-33
Supplier:
JENWAY
Expand image
Accessories for Jenway meters
Catalog # JENW99960-33
Supplier:
JENWAY
Specifications
Description:
Electrode stand and holder
Pk:
1
Specifications
About this item
Product Details
Product Details