Thymolphthalein ACS, Reag. Ph. Eur. indicator
Supplier: Merck
Certificates
About this item
CAS Number:125-20-2
:
Formula:C₂₈H₃₀O₄
MDL Number:MFCD00005909
:430.54 g/mol
Merck Index:12,09542
Melting Pt:253 °C
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:204-729-7
Specification Test Results
- Appearance:White to almost white powder, eventually slightly yellowish.
- Identity (UV/VIS-Spectrum):passes test
- Absorption maximum λmax. (buffer pH 10.5):592 - 596 nm
- Spec. Absorptivity A 1%/1cm (λmax; 0.01 g/l; buffer pH 10.5; calculated on anhydrous substance):800 - 900
- Transition range (according to ACS):passes test
- Transition range:pH 9.0 - pH 10.5 colourless - blue
- Loss on drying (110 °C):≤ 1 %
Specification Test Results
Related Information
Frequently Bought Together