Order Entry
Slovakia
ContactUsLinkComponent
EGTA (ethylene glycol bis(2-aminoethyl ether)-N,N,N',N'-tetraacetic acid) ≥97%, white crystalline powder, Ultra Pure Grade
EGTA (ethylene glycol bis(2-aminoethyl ether)-N,N,N',N'-tetraacetic acid) ≥97%, white crystalline powder, Ultra Pure Grade
Supplier:  VWR Chemicals
Certificates
undefined
EGTA (ethylene glycol bis(2-aminoethyl ether)-N,N,N',N'-tetraacetic acid) ≥97%, white crystalline powder, Ultra Pure Grade
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • DNase:
    none detected
  • Heavy Metals (as Pb):
    ≤0.001%
  • Loss on Drying:
    ≤1%
  • Melting Range w/Decomposition:
    ≥238 °C
  • Protease:
    none
  • Purity:
    >97 %
  • RNase:
    none detected

Specification Test Results

Related Information

Frequently Bought Together