- Description:EZ-Pak® Membrane dispenser curve
EZ-Pak® Membrane dispenser curve has robust components selected for durability and high-throughput dispensing. New design with cover allows you to load a cartridge in less than 30 seconds. Membrane is consistently well-positioned for easy transfer with forceps.
- High-speed sterile membrane dispensing
- No-touch operation
- One-handed membrane transfer
- The smooth shape facilitates cleaning
- The plastic material is compatible with a large range of disinfection agents
- Lightweight and portable
No need to press a lever, the durable motor will dispense one membrane in less than one second. Move your hand toward the sensor - no touch necessary and a membrane is unwrapped, dispensed and automatically positioned for transfer with forceps. Software controls prevent the accidental dispensing of multiple membranes.
The dispenser can work with or without connection to a power outlet. When the Lithium ion battery
is fully charged, more than 10000 membranes can be dispensed. After 10 minutes idle time, a standby mode is activated to save energy.