Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovakia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Potassium hydroxide
Potassium hydroxide 2 mol/l (2 N) in ethanol 90% Reag. Ph. Eur. 1070301
Potassium hydroxide 2 mol/l (2 N) in ethanol 90% Reag. Ph. Eur. 1070301
Potassium hydroxide 2 mol/l (2 N) in ethanol 90% Reag. Ph. Eur. 1070301
87904.180
:
VWR Chemicals
:
Potassium hydroxide 2 mol/l (2 N) in ethanol 90% Reag. Ph. Eur. 1070301
87904.180
:
VWR Chemicals
:
Pk:
100 mL
Seq:
2440
:
:
56.11 g/mol
:
Ambient
:
215-181-3
:
2924
:
3,II
:
GHS02
:
GHS05
:
1310-58-3
:
KOH
:
MFCD00003553