Supelco® PESTANAL® Organic Reference Standard, β-Endosulfan
33385-100MG
: Merck
:
- Pack type:Glass bottle for solids
- Pk:100 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:33213-65-9
- Description:β-Endosulfan
- Storage conditions:2...30 °C
- Hazard class description:H301 - H310 + H330 - H410
- Shipping conditions:STD
- MDL No.:MFCD00151176
- M-Clarity quality level:MQ100
- Molecular weight:406,93
- Flash point:Not applicable
- GHS signal: GHS06, GHS09
: P260 - P262 - P273 - P280 - P302 + P352 + P310 - P304 + P340 + P310