Order Entry
Slovakia
ContactUsLinkComponent
Supelco® PESTANAL® Organic Reference Standard, α-Endosulfan
Supelco® PESTANAL® Organic Reference Standard, α-Endosulfan
  45468-100MG
 :  Merck
 :  
undefined
Supelco® PESTANAL® Organic Reference Standard, α-Endosulfan
  45468-100MG
 :  Merck
 :  

 

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 mg
  • Standards:
    PESTANAL®, Analytical standard
  • CAS No.:
    959-98-8
  • Description:
    α-Endosulfan
  • Storage conditions:
    2...30 °C
  • Hazard class description:
    H300 + H330 - H312 - H410
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00151174
  • M-Clarity quality level:
    MQ100
  • Molecular weight:
    406,93
  • Flash point:
    Not applicable

 

 

  • GHS signal: GHS06, GHS09
 : P273 - P280 - P301 + P310 + P330 - P302 + P352 + P312 - P304 + P340 + P310