1-Pentanesulphonic acid sodium salt, LiChropur™ for ion pair chromatography, Supelco®
1.18304.0025
: Merck
:
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
- Hill Formula:C₅H₁₁NaO₃S
- Molar Mass:174.20 g/mol
- pH value:5.5 - 7.5 (100 g/l, H₂O, 20 °C)
- Bulk density:390 kg/m³
- Solubility:(20 °C) soluble
- Storage class:10 - 13 Other liquids and solids
- WGK:WGK 3 highly water endangering
:
:174.2 g/mol
:> 300 °C
:M-Clarity™ quality level = MQ100
:Ambient
:245-208-4
:22767-49-3
:C₅H₁₁O₃SNa
:MFCD00007541