Order Entry
Slovakia
Orders LinkContactUsLinkComponent
 

Barium aluminate, tech.

Barium aluminate, tech.
Certificates
Barium aluminate, tech.
Barium aluminate, tech.

2 Options Available Below

Product Details & Documents

CAS No.: 12004-04-5 Molecular Formula: BaAl2O4 (approximate formula) Formula Weight: 561.98 Physical Form: Powder Odour: Odourless UN No.: 1564 MDL No.: MFCD00049716

  • CAS Number:12004-04-5
  • Formula:Al₂BaO₄
  • MDL Number:MFCD00049716
  • Molecular Weight:255.29 g/mol
  • Melting Pt:169.5…170 °C
  • Density:4.04 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:234-445-9
  • UN:1564
  • ADR:6.1,III
Product Family Options

Product Information

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.