Human Serum Albumin (HSA) Powder
Supplier: GEMINI BIO-PRODUCTS
New Product
Certificates
About this item
Human albumin is prepared by cold-alcohol fractionation from pooled human plasma and lyophilised. MW ~66 kDa.
Caution: For cell culture, further manufacturing and research use only. Not for direct therapeutic use.
Specifications
- Catalog No:
- GEMB800-125P-025
- GEMB800-125P-005
- GEMB800-125P-001
- Shipping conditions:
- Ships ambient
- Ships ambient
- Ships ambient
- Storage temperature:
- Store refrigerated at 2...8 °C
- Store refrigerated at 2...8 °C
- Store refrigerated at 2...8 °C
Specifications
Frequently Bought Together